Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmbr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029098.3 | NP_083374.1 | 17 | 2131 | ||
GenBank mRNA | AK003656.1 | - | 17 | 2063 | ||
AK030589.1 | - | 17 | 2097 | |||
AK049110.1 | - | 17 | 2132 | |||
AK143891.1 | - | 17 | 2092 | |||
AK154968.1 | - | 17 | 2265 | |||
AK173121.1 | - | 17 | 2512 | |||
BC023397.1 | - | 17 | 2068 | 5043877 |