Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029101.4 | NP_083377.3 | 7 | 1830 | ||
GenBank mRNA | AK003698.1 | - | 7 | 1781 | ||
AK032596.1 | - | 1793 | ||||
AK036247.1 | - | 1775 | ||||
AK088456.1 | - | 7 | 1809 | |||
AK146821.1 | - | 7 | 1793 | |||
AK153565.1 | - | 7 | 1789 | |||
AK159553.1 | - | 7 | 1775 | |||
AK168189.1 | - | 8 | 1767 | |||
BC012523.1 | - | 7 | 1767 | 3602183 |