Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006407.3 | NP_006398.1 | 2 | 469 | ||
GenBank mRNA | AB014765.1 | - | 2 | 375 | ||
AB052638.1 | - | 2 | 367 | |||
AB097051.1 | - | 2 | 450 | |||
AF070523.1 | - | 2 | 475 | |||
AF115965.1 | - | 2 | 460 | |||
AF125530.1 | - | 2 | 461 | |||
AF161476.1 | - | 2 | 450 | |||
AK074612.1 | - | 2 | 421 | |||
AK223566.1 | - | 2 | 373 | |||
AK301637.1 | - | 3 | 383 | |||
AK303148.1 | - | 2 | 355 | |||
AK312731.1 | - | 2 | 372 | |||
AY102608.1 | - | 2 | 360 | |||
BC005143.2 | - | 2 | 392 | 3510436 | ||
BC020797.1 | - | 2 | 386 | 4134658 |