Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eppin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029325.2 | NP_083601.1 | 3, 4 | 455 | ||
GenBank mRNA | AF346413.2 | - | 3, 4 | 478 | ||
AK006296.1 | - | 3, 4 | 457 | |||
BC048637.1 | - | 3, 4 | 478 | 6771674 |