Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006277.2 | NP_006268.2 | 12 | 1585 | ||
NM_019595.3 | NP_062541.3 | 12 | 1585 | |||
NM_147152.2 | NP_671494.2 | 12 | 1585 | |||
GenBank mRNA | AB033082.1 | - | 12 | 1492 | ||
AF182198.1 | - | 12 | 1564 | |||
AF182199.1 | - | 12 | 1564 | |||
AF248540.1 | - | 11 | 1366 | |||
AK021545.1 | - | 5 | 573 | |||
BC020921.2 | - | 11 | 1362 | |||
BC038963.1 | - | 13 | 1651 | |||
BC146779.1 | - | 12 | 1492 | 9007231 |