Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290679.1 | NP_001277608.1 | 5 | 877 | ||
NM_029398.3 | NP_083674.2 | 5 | 694 | |||
GenBank mRNA | AK017734.1 | - | 5 | 791 | ||
BC034768.1 | - | 5 | 640 | 4039224 | ||
BC060034.1 | - | 5 | 638 | 6491675 |