Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285959.1 | NP_001272888.1 | 4 | 440 | ||
NM_001285960.1 | NP_001272889.1 | 4 | 573 | |||
NM_029610.3 | NP_083886.1 | 4 | 824 | |||
GenBank mRNA | AK009135.1 | - | 4 | 827 | ||
BC011339.1 | - | 4 | 614 | 3978191 | ||
DV043140.1 | - | 3 | 598 |