Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grhpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080289.1 | NP_525028.1 | 9 | 1020 | ||
GenBank mRNA | AK004307.1 | - | 9 | 1036 | ||
AK018293.1 | - | 9 | 1035 | |||
AK168351.1 | - | 9 | 1017 | |||
AK172201.1 | - | 9 | 988 | |||
AY113690.1 | - | 9 | 994 | |||
BC010194.1 | - | 9 | 1020 | 3966541 |