Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088789.1 | - | 13 | 2793 | ||
AK145003.1 | - | 13 | 2782 | |||
BC004596.1 | - | 13 | 2740 | 3584555 | ||
BC004665.1 | - | 3 | 1414 |