Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133743.1 | NP_598504.1 | 5 | 1255 | ||
GenBank mRNA | AK009998.1 | - | 5 | 1275 | ||
BC003749.1 | - | 5 | 1257 | |||
BC016549.1 | - | 5 | 1255 | 3585545 | ||
BC094282.1 | - | 5 | 1292 | 5029753 |