Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286007.1 | NP_001272936.1 | 6 | 1146 | ||
NM_001286009.1 | NP_001272938.1 | 6 | 1146 | |||
NM_001286011.1 | NP_001272940.1 | 5 | 913 | |||
NM_133755.2 | NP_598516.2 | 6 | 1073 | |||
GenBank mRNA | AK033157.1 | - | 6 | 863 | ||
AK045084.1 | - | 6 | 862 | |||
AK146771.1 | - | 6 | 1068 | |||
AK153511.1 | - | 6 | 994 | |||
AK158060.1 | - | 6 | 862 | |||
AK167924.1 | - | 6 | 968 | |||
BC012519.1 | - | 6 | 1001 | 3601589 | ||
BC025564.1 | - | 5 | 835 | |||
BC025582.1 | - | 6 | 962 | 5324981 | ||
BC027034.1 | - | 6 | 1057 |