Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144533.2 | NP_653116.1 | 5 | 878 | ||
GenBank mRNA | AK041897.1 | - | 4 | 653 | ||
BC005737.1 | - | 5 | 878 | 3599134 | ||
BC092086.1 | - | 5 | 587 | 5040019 |