Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278196.1 | NP_001265125.1 | 2 | 825 | ||
NM_006352.4 | NP_006343.2 | 1 | 1353 | |||
NM_205768.2 | NP_991331.1 | 2 | 956 | |||
XM_005273006.2 | XP_005273063.1 | 3 | 886 | |||
XM_017000060.1 | XP_016855549.1 | 3 | 2986 | |||
GenBank mRNA | AJ001388.1 | - | 1 | 1279 | ||
AK291408.1 | - | 2 | 933 | |||
AK293892.1 | - | 2 | 935 | |||
BC036677.2 | - | 2 | 932 | 5259205 | ||
BC078175.1 | - | 1 | 1353 | |||
X95072.1 | - | 1 | 1072 |