Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175168.4 | NP_780377.1 | 20 | 3984 | ||
GenBank mRNA | AB055408.1 | - | 6 | 1483 | ||
AK018379.1 | - | 2 | 828 | |||
AK053044.1 | - | 20 | 3917 | |||
AK163135.1 | - | 20 | 3917 | |||
BC027800.1 | - | 9 | 2039 | |||
BC037175.1 | - | 5 | 2579 | |||
BC076578.1 | - | 20 | 3984 | 30619280 |