Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175224.4 | NP_780433.1 | 11 | 1627 | ||
GenBank mRNA | AK077694.1 | - | 11 | 1628 | ||
AK129054.1 | - | 6 | 3734 | |||
AK153944.1 | - | 11 | 1634 | |||
BC027110.1 | - | 6 | 1167 | |||
BC096469.1 | - | 11 | 1616 | 3603051 |