Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025105.2 | NP_001020276.1 | 10, 11 | 1686 | ||
NM_001271742.1 | NP_001258671.1 | 9, 10 | 1125 | |||
NM_001892.5 | NP_001883.4 | 9, 10 | 1602 | |||
GenBank mRNA | AK293761.1 | - | 9, 10 | 1092 | ||
AK303794.1 | - | 10, 11 | 1134 | |||
BC008717.2 | - | 9, 10 | 1144 | 2822250 | ||
L37042.1 | - | 9, 10 | 1225 |