Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176860.5 | NP_789830.1 | 12 | 1686 | ||
GenBank mRNA | AB075602.1 | - | 12 | 1681 | ||
AK013361.1 | - | 12 | 1754 | |||
AK034450.1 | - | 12 | 1877 | |||
AK035764.1 | - | 12 | 1687 | |||
AK133948.1 | - | 12 | 1684 | |||
AK150909.1 | - | 12 | 2253 | |||
AK151721.1 | - | 12 | 2254 | |||
AK151895.1 | - | 12 | 1887 | |||
AK152013.1 | - | 12 | 1887 | |||
AK154576.1 | - | 12 | 1908 | |||
BC053436.1 | - | 12 | 1658 | 6510702 |