Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177323.3 | NP_796297.2 | 2 | 173 | ||
GenBank mRNA | AK005284.1 | - | 2 | 206 | ||
AK013321.1 | - | 1 | 606 | |||
AK028707.1 | - | 2 | 204 | |||
AK034857.1 | - | 1 | 604 | |||
AK133789.1 | - | 2 | 199 | |||
AK135334.1 | - | 2 | 194 | |||
AK136479.1 | - | 2 | 179 | |||
AK144144.1 | - | 2 | 195 | |||
AK160710.1 | - | 2 | 173 | |||
AK162664.1 | - | 2 | 150 | |||
BC065059.1 | - | 2 | 173 | 6401664 | ||
BC145267.1 | - | 2 | 163 | 9053821 | ||
BC150745.1 | - | 2 | 163 | 9087656 |