Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178398.4 | NP_848485.1 | 2 | 318 | ||
GenBank mRNA | AK010881.1 | - | 2 | 346 | ||
AK155113.1 | - | 2 | 348 | |||
AK169051.1 | - | 2 | 292 | |||
AK172030.1 | - | 2 | 294 | |||
AK194559.1 | - | 1 | 47 | |||
BC044894.1 | - | 2 | 343 | 4235237 |