Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urgcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077661.1 | NP_001071129.1 | 6 | 1861 | ||
NM_178623.3 | NP_848738.2 | 6 | 1445 | |||
GenBank mRNA | AK033577.1 | - | 6 | 1419 | ||
AK035580.1 | - | 6 | 1857 | |||
AK041953.1 | - | 7 | 2110 | |||
AK129378.1 | - | 5 | 1503 | |||
AK166495.1 | - | 5 | 1345 | |||
BC066994.1 | - | 6 | 1742 | 5696000 |