Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178628.5 | NP_848743.1 | 5, 6 | 914 | ||
GenBank mRNA | AK046919.1 | - | 4, 5 | 644 | ||
AK050339.1 | - | 5, 6 | 751 | |||
AK140185.1 | - | 4, 5 | 658 | |||
BC050784.1 | - | 4, 5 | 643 | 6595181 |