Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097590.2 | NP_001091059.1 | 3 | 484 | ||
NM_001265601.1 | NP_001252530.1 | 2 | 722 | |||
NM_006414.4 | NP_006405.2 | 2 | 365 | |||
NM_183005.4 | NP_892117.1 | 3 | 841 | |||
XM_006717363.1 | XP_006717426.1 | 3 | 655 | |||
XM_006717364.3 | XP_006717427.1 | 2 | 277 | |||
XM_011519293.1 | XP_011517595.1 | 2 | 520 | |||
XM_017015479.1 | XP_016870968.1 | 3 | 973 | |||
XM_017015480.1 | XP_016870969.1 | 3 | 431 | |||
XM_017015481.1 | XP_016870970.1 | 2 | 455 | |||
GenBank mRNA | AK057006.1 | - | 3 | 639 | ||
AK223418.1 | - | 2 | 342 | |||
BC029494.1 | - | 3 | 622 | 5271044 | ||
BP247818.1 | - | 2 | 362 | |||
BU180901.1 | - | 2 | 482 | |||
BU197354.1 | - | 3 | 507 | |||
CR541816.1 | - | 1 | 125 | |||
CX787861.1 | - | 2 | 754 | |||
CX872398.1 | - | 3 | 416 | |||
U77664.1 | - | 2 | 161 |