Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4921517D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183290.2 | NP_899113.1 | 4 | 591 | ||
GenBank mRNA | AK014909.1 | - | 4 | 592 | ||
AK133255.1 | - | 4 | 599 | |||
BC147600.1 | - | 3 | 380 | 9056590 | ||
BC147624.1 | - | 3 | 380 | 9056614 |