Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006418.4 | NP_006409.3 | 3, 4 | 668 | ||
GenBank mRNA | AF097021.1 | - | 3, 4 | 585 | ||
AK300947.1 | - | 3, 4 | 548 | |||
AY358567.1 | - | 3, 4 | 585 | |||
BC047740.1 | - | 2, 3 | 299 | |||
BC117329.1 | - | 3, 4 | 650 | 40125880 |