Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022885.2 | NP_075023.2 | 13 | 1837 | ||
GenBank mRNA | AF233321.1 | - | 6 | 856 | ||
AF461761.1 | - | 13 | 1736 | |||
AK007424.1 | - | 4 | 357 | |||
AK051183.1 | - | 13 | 1837 | |||
AK088004.1 | - | 12 | 1581 | |||
AK090176.1 | - | 13 | 1746 | |||
AK166006.1 | - | 13 | 1647 | |||
AK187253.1 | - | 2 | 50 | |||
BC029033.1 | - | 10 | 1279 | |||
BC029217.1 | - | 6 | 689 | |||
BC033452.1 | - | 13 | 1736 | 3156622 |