Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adck3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163290.1 | NP_001156762.1 | 15 | 1803 | ||
NM_023341.3 | NP_075830.2 | 15 | 1901 | |||
GenBank mRNA | AJ278735.1 | - | 15 | 1781 | ||
AK004791.1 | - | 15 | 1843 | |||
AK014605.2 | - | 15 | 1843 | |||
AK197823.1 | - | 2 | 110 | |||
BC030937.1 | - | 15 | 1849 | 4922892 |