Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023852.5 | NP_076341.1 | 5 | 926 | ||
GenBank mRNA | AK014132.1 | - | 5 | 848 | ||
AK079574.1 | - | 5 | 823 | |||
AK148904.1 | - | 5 | 847 | |||
AK161006.1 | - | 5 | 824 | |||
AK164316.1 | - | 5 | 871 | |||
AY026947.1 | - | 5 | 807 | |||
BC114335.2 | - | 5 | 799 | 40045744 |