Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024214.2 | NP_077176.1 | 4, 5 | 588 | ||
GenBank mRNA | AK002902.1 | - | 4, 5 | 509 | ||
AK160168.1 | - | 4, 5 | 491 | |||
AK218667.1 | - | 1, 2 | 48 | |||
BC002087.1 | - | 4, 5 | 497 | 3484983 | ||
BC099449.1 | - | 4, 5 | 495 | 30848526 |