Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERLIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100626.1 | NP_001094096.1 | 3 | 233 | ||
NM_006459.3 | NP_006450.2 | 2 | 477 | |||
XM_005269442.3 | XP_005269499.1 | 3 | 263 | |||
XM_017015541.1 | XP_016871030.1 | 3 | 247 | |||
GenBank mRNA | AF064093.1 | - | 3 | 209 | ||
AK056574.1 | - | 2 | 2834 | |||
AK222480.1 | - | 3 | 205 | |||
AK315580.1 | - | 3 | 205 | |||
BC031791.1 | - | 2 | 146 | 4214940 | ||
BP280488.1 | - | 3 | 233 | |||
BU192550.1 | - | 2 | 477 |