Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026058.4 | NP_080334.3 | 6, 7 | 716 | ||
GenBank mRNA | AK013554.1 | - | 6, 7 | 668 | ||
AK028937.1 | - | 6, 7 | 642 | |||
AK028945.1 | - | 6, 7 | 619 | |||
AK030280.1 | - | 6, 7 | 656 | |||
AK036108.1 | - | 6 | 1342 | |||
AK134887.1 | - | 6 | 1375 | |||
AK215994.1 | - | 3, 4 | 169 | |||
AY029531.1 | - | 6, 7 | 617 | |||
BC003946.1 | - | 6, 7 | 603 | 3487343 |