Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026058.4 | NP_080334.3 | 11 | 1412 | ||
GenBank mRNA | AK013554.1 | - | 11 | 1364 | ||
AK021316.1 | - | 4 | 519 | |||
AK028937.1 | - | 11 | 1338 | |||
AK028945.1 | - | 11 | 1315 | |||
AK030280.1 | - | 11 | 1352 | |||
AK076409.1 | - | 2 | 346 | |||
AK134887.1 | - | 10 | 2071 | |||
AK201349.1 | - | 1 | 14 | |||
AK210387.1 | - | 2 | 366 | |||
AY029531.1 | - | 11 | 1313 | |||
BC003946.1 | - | 11 | 1299 | 3487343 |