Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pbld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026085.2 | NP_080361.1 | 10 | 2074 | ||
GenBank mRNA | AK014198.1 | - | 10 | 2075 | ||
AK018692.1 | - | 9 | 1830 | |||
BC061073.1 | - | 9 | 1829 | 30381600 |