Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem167b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026198.2 | NP_080474.2 | 3 | 1115 | ||
GenBank mRNA | AK008415.1 | - | 3 | 1028 | ||
AK046697.1 | - | 3 | 1016 | |||
AK155399.1 | - | 3 | 1011 | |||
BC050932.1 | - | 3 | 1019 | 6811263 | ||
BC055449.1 | - | 3 | 1032 | 3993993 |