Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026223.2 | NP_080499.1 | 1, 2 | 243 | ||
GenBank mRNA | AK014846.1 | - | 1, 2 | 244 | ||
AK015834.1 | - | 1, 2 | 227 | |||
AK161292.1 | - | 1, 2 | 244 | |||
BC118017.1 | - | 1, 2 | 244 | 40102576 |