Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026235.4 | NP_080511.2 | 2 | 378 | ||
GenBank mRNA | AK017372.1 | - | 2 | 383 | ||
AK020966.1 | - | 2 | 315 | |||
AK039614.1 | - | 2 | 361 | |||
AK042616.1 | - | 2 | 310 | |||
AK087521.1 | - | 2 | 381 | |||
BC090615.1 | - | 2 | 510 | 6809572 |