Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026239.2 | NP_080515.1 | 2 | 417 | ||
GenBank mRNA | AK005180.1 | - | 2 | 252 | ||
AK018535.1 | - | 2 | 254 | |||
AK042538.1 | - | 2 | 253 | |||
AK043881.1 | - | 2 | 252 | |||
BC042623.1 | - | 2 | 417 | 5369317 | ||
BC050880.1 | - | 2 | 319 | 5690508 |