Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026242.3 | NP_080518.1 | 2 | 629 | ||
GenBank mRNA | AB041651.1 | - | 2 | 611 | ||
AK005313.1 | - | 2 | 617 | |||
AK009827.1 | - | 2 | 584 | |||
AK017832.1 | - | 2 | 611 | |||
AK018675.1 | - | 2 | 580 | |||
AK159464.1 | - | 2 | 617 | |||
AK167112.1 | - | 2 | 611 | |||
AK170789.1 | - | 2 | 580 | |||
AK170909.1 | - | 2 | 606 | |||
AK170984.1 | - | 2 | 612 | |||
BC010209.1 | - | 2 | 598 | 3482091 | ||
BC085488.1 | - | 2 | 474 |