Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026242.3 | NP_080518.1 | 2 | 1004 | ||
GenBank mRNA | AB041651.1 | - | 2 | 984 | ||
AK005313.1 | - | 2 | 996 | |||
AK009827.1 | - | 2 | 962 | |||
AK017832.1 | - | 2 | 986 | |||
AK018675.1 | - | 2 | 954 | |||
AK020049.1 | - | 2 | 972 | |||
AK159464.1 | - | 2 | 993 | |||
AK167112.1 | - | 2 | 985 | |||
AK170789.1 | - | 2 | 954 | |||
AK170909.1 | - | 2 | 982 | |||
AK170984.1 | - | 2 | 986 | |||
AK217266.1 | - | 1 | 127 | |||
BC010209.1 | - | 2 | 973 | 3482091 | ||
BC085488.1 | - | 2 | 849 |