Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026242.3 | NP_080518.1 | 2 | 754 | ||
GenBank mRNA | AB041651.1 | - | 2 | 734 | ||
AK005313.1 | - | 2 | 746 | |||
AK009827.1 | - | 2 | 712 | |||
AK018675.1 | - | 2 | 704 | |||
AK020049.1 | - | 2 | 722 | |||
AK159464.1 | - | 2 | 743 | |||
AK167112.1 | - | 2 | 736 | |||
AK170789.1 | - | 2 | 704 | |||
AK170909.1 | - | 2 | 732 | |||
AK170984.1 | - | 2 | 736 | |||
BC010209.1 | - | 2 | 723 | 3482091 | ||
BC085488.1 | - | 2 | 599 |