Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306075.1 | NP_001293004.1 | 2 | 698 | ||
NM_006527.3 | NP_006518.1 | 3 | 815 | |||
GenBank mRNA | AK091735.1 | - | 2 | 275 | ||
AK093314.1 | - | 3 | 288 | |||
AK094257.1 | - | 3 | 629 | |||
AK315635.1 | - | 3 | 386 | |||
BC014908.1 | - | 3 | 384 | 3908584 | ||
BC015703.1 | - | 3 | 384 | 3907984 | ||
BT007433.1 | - | 3 | 262 | |||
DB027411.1 | - | 3 | 422 | |||
DB031118.1 | - | 3 | 288 | |||
U75679.1 | - | 3 | 377 | |||
Z71188.1 | - | 3 | 366 |