Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026278.3 | NP_080554.1 | 5 | 562 | ||
GenBank mRNA | AK007202.1 | - | 5 | 554 | ||
AK016626.1 | - | 567 | ||||
BC030908.1 | - | 3 | 182 | |||
BC051150.1 | - | 5 | 562 | 6772814 |