Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006537.3 | NP_006528.2 | 2, 3 | 389 | ||
NR_046341.1 | 3, 4 | 542 | ||||
XM_017022763.1 | XP_016878252.1 | 4, 5 | 1388 | |||
XM_017022764.1 | XP_016878253.1 | 3, 4 | 1324 | |||
XM_017022765.1 | XP_016878254.1 | 3, 4 | 530 | |||
GenBank mRNA | AF073344.1 | - | 2, 3 | 253 | ||
AK300948.1 | - | 2, 3 | 244 | |||
AK307810.1 | - | 2, 3 | 147 | |||
AK309382.1 | - | 1, 2 | 210 | |||
AK314223.1 | - | 2, 3 | 308 | |||
AY461579.1 | - | 3, 4 | 418 | |||
BC018113.2 | - | 2, 3 | 254 | 3916811 | ||
BC065300.1 | - | 2, 3 | 289 | 6146202 | ||
BC100029.1 | - | 2, 3 | 261 | |||
BC107137.1 | - | 2, 3 | 249 | 40025968 | ||
BC107138.1 | - | 2, 3 | 249 | 40025969 | ||
BT007269.1 | - | 2, 3 | 160 | |||
DA557812.1 | - | 2, 3 | 390 |