Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026423.4 | NP_080699.3 | 4 | 816 | ||
GenBank mRNA | AK007709.1 | - | 4 | 777 | ||
AK010538.1 | - | 4 | 788 | |||
AK013882.1 | - | 4 | 785 | |||
AK047287.1 | - | 4 | 819 | |||
AK144807.1 | - | 4 | 759 | |||
AK164894.1 | - | 4 | 785 | |||
AK177142.1 | - | 1 | 40 | |||
AK185486.1 | - | 2 | 125 | |||
BC025049.1 | - | 4 | 783 | 4951045 |