Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006421.4 | NP_006412.2 | 1, 2 | 524 | ||
XM_005251134.4 | XP_005251191.1 | 2, 3 | 503 | |||
XM_005251135.3 | XP_005251192.1 | 1, 2 | 774 | |||
XM_005251136.3 | XP_005251193.1 | 1, 2 | 774 | |||
XM_011517442.2 | XP_011515744.1 | 2, 3 | 810 | |||
GenBank mRNA | AF084520.1 | - | 1, 2 | 134 | ||
AF111162.1 | - | 1, 2 | 275 | |||
AI523655.1 | - | 2, 3 | 351 |