Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026470.3 | NP_080746.3 | 13 | 1675 | ||
GenBank mRNA | AF032967.1 | - | 7 | 982 | ||
AK005819.1 | - | 8 | 1062 | |||
AK006861.1 | - | 8 | 1047 | |||
AK132771.1 | - | 13 | 1649 | |||
AK153098.1 | - | 13 | 1552 | |||
AK212466.1 | - | 1 | 180 |