Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdhaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026503.3 | NP_080779.2 | 3 | 548 | ||
GenBank mRNA | AK003789.1 | - | 3 | 506 | ||
AK004301.1 | - | 3 | 528 | |||
AK007909.1 | - | 3 | 541 | |||
AK015824.1 | - | 3 | 549 | |||
BC028981.1 | - | 3 | 513 | 5367409 |