Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168471.1 | NP_001161943.1 | 3 | 1625 | ||
NM_001168472.1 | NP_001161944.1 | 3 | 1798 | |||
NM_026556.4 | NP_080832.1 | 3 | 1831 | |||
GenBank mRNA | AK028319.1 | - | 3 | 1596 | ||
AK049645.1 | - | 3 | 1805 | |||
AK160114.1 | - | 3 | 1801 | |||
AK169212.1 | - | 3 | 1805 | |||
BC011289.1 | - | 3 | 1618 | 3981362 | ||
BC040822.1 | - | 3 | 1789 | 4459858 | ||
BC058797.1 | - | 3 | 1437 |