Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026578.3 | NP_080854.1 | 7 | 1117 | ||
GenBank mRNA | AK021256.1 | - | 7 | 878 | ||
AK075923.1 | - | 6 | 852 | |||
AK167001.1 | - | 7 | 857 | |||
BC021873.1 | - | 8 | 981 | 3709271 | ||
BC048685.1 | - | 7 | 1036 | 6510731 |