Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026626.2 | NP_080902.1 | 2 | 299 | ||
GenBank mRNA | AK010880.1 | - | 2 | 152 | ||
AK015157.1 | - | 2 | 81 | |||
AK018894.1 | - | 2 | 160 | |||
AK019663.1 | - | 2 | 92 | |||
AK019802.1 | - | 2 | 101 | |||
AK029880.1 | - | 82 | ||||
BC025062.1 | - | 2 | 299 | 4240831 |