Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026645.3 | NP_080921.2 | 8 | 692 | ||
GenBank mRNA | AK016688.1 | - | 8 | 693 | ||
AK018835.1 | - | 8 | 695 | |||
AK132739.1 | - | 8 | 697 | |||
BC049549.1 | - | 8 | 693 | 6774389 | ||
DQ153247.1 | - | 8 | 698 |